2-Bromo-1-ethoxy-3-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

152355-1G 152355-500MG 152355-5G 152355-25G
77465-074EA 415.09 USD
77465-074 77477-842 77487-980 77498-672
2-Bromo-1-ethoxy-3-fluoro-4-methoxybenzene ≥97%
2-Bromo-1-ethoxy-3-fluoro-4-methoxybenzene

Precaución: For research use only

Formula: C₉H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34842770
CAS Number: 2383596-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR