2-Fluoro-4-methyl-3-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

152090-1G 152090-500MG 152090-5G 152090-25G
77465-070EA 415.09 USD
77465-070 77477-838 77487-976 77498-666
2-Fluoro-4-methyl-3-phenylpyridine ≥97%
2-Fluoro-4-methyl-3-phenylpyridine

Precaución: For research use only

Formula: C₁₂H₁₀FN
MW: 187.213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701873
CAS Number: 1214388-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR