4-Bromo-2-(methylthio)benzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

151305-1G 151305-500MG 151305-5G 151305-25G
77465-062EA 415.09 USD
77465-062 77477-830 77487-966 77498-654
4-Bromo-2-(methylthio)benzo[b]thiophene ≥95%
4-Bromo-2-(methylthio)benzo[b]thiophene

Precaución: For research use only

Formula: C₉H₇BrS₂
Storage Temperature: Ambient
MDL Number: MFCD34842167
CAS Number: 2921895-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR