4-Bromo-1-benzothiophene-2-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

151302-1G 151302-500MG 151302-5G 151302-25G
77465-058EA 199.82 USD
77465-058 77477-826 77487-962 77498-652
4-Bromo-1-benzothiophene-2-carbaldehyde ≥95%
4-Bromo-1-benzothiophene-2-carbaldehyde

Precaución: For research use only

Formula: C₉H₅BrOS
Storage Temperature: Ambient
MDL Number: MFCD19381762
CAS Number: 19075-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR