(2,6-Dimethoxy-4-(trifluoromethyl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

15121-1G 15121-500MG 15121-5G 15121-25G
77465-054EA 415.09 USD
77465-054 77477-822 77487-958 77498-650
(2,6-Dimethoxy-4-(trifluoromethyl)phenyl)boronic acid ≥97%
(2,6-Dimethoxy-4-(trifluoromethyl)phenyl)boronic acid

Precaución: For research use only

Formula: C₉H₁₀BF₃O₄
Storage Temperature: Ambient
MDL Number: MFCD34755679
CAS Number: 232275-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR