2-Chloro-1-(3-chloro-2,6-difluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150082-1G 150082-500MG 150082-5G 150082-25G
77465-046EA 415.09 USD
77465-046 77477-814 77487-950 77498-644
2-Chloro-1-(3-chloro-2,6-difluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3-chloro-2,6-difluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂Cl₂F₄O
Storage Temperature: Ambient
MDL Number: MFCD34841212
CAS Number: 2921908-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR