2-Chloro-1-(2,6-difluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150078-1G 150078-500MG 150078-5G
77465-042EA 594.31 USD
77465-042 77477-810 77487-946
2-Chloro-1-(2,6-difluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,6-difluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₃ClF₄O
MW: 226.555 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045713
CAS Number: 1564523-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR