2-Chloro-1-(6-chloro-2-fluoro-3-methoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150069-1G 150069-500MG 150069-5G 150069-25G
77465-038EA 415.09 USD
77465-038 77477-806 77487-942 77498-640
2-Chloro-1-(6-chloro-2-fluoro-3-methoxyphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(6-chloro-2-fluoro-3-methoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅Cl₂F₃O₂
MW: 273.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841202
CAS Number: 2921861-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR