1-(2-Bromo-6-fluoro-4-methylphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150060-1G 150060-500MG 150060-5G 150060-25G
77465-034EA 415.09 USD
77465-034 77477-802 77487-938 77498-636
1-(2-Bromo-6-fluoro-4-methylphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(2-Bromo-6-fluoro-4-methylphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD34841193
CAS Number: 2921908-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR