1-(3-Bromo-2-chloro-6-fluorophenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150056-1G 150056-500MG 150056-5G 150056-25G
77465-030EA 415.09 USD
77465-030 77477-798 77487-934 77498-632
1-(3-Bromo-2-chloro-6-fluorophenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(3-Bromo-2-chloro-6-fluorophenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂BrCl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD34841189
CAS Number: 2921879-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR