1-(6-Bromo-2-fluoro-3-methylphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150053-1G 150053-500MG 150053-5G 150053-25G
77465-026EA 415.09 USD
77465-026 77477-794 77487-930 77498-628
1-(6-Bromo-2-fluoro-3-methylphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(6-Bromo-2-fluoro-3-methylphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O
MW: 301.488 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841186
CAS Number: 2921865-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR