1-(5-Bromo-3-ethoxy-2-fluorophenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150038-1G 150038-500MG 150038-5G 150038-25G
77465-022EA 415.09 USD
77465-022 77477-790 77487-926 77498-624
1-(5-Bromo-3-ethoxy-2-fluorophenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(5-Bromo-3-ethoxy-2-fluorophenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₇BrClF₃O₂
MW: 331.514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841172
CAS Number: 2921894-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR