2-Chloro-2,2-difluoro-1-(2-fluoro-5-iodophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

150019-1G 150019-500MG 150019-5G 150019-25G
77465-018EA 415.09 USD
77465-018 77477-786 77487-922 77498-620
2-Chloro-2,2-difluoro-1-(2-fluoro-5-iodophenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-fluoro-5-iodophenyl)ethanone

Precaución: For research use only

Formula: C₈H₃ClF₃IO
MW: 334.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841153
CAS Number: 2921729-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR