2-Chloro-2,2-difluoro-1-(2-fluoro-5-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

150004-1G 150004-500MG 150004-5G
77465-014EA 546.93 USD
77465-014 77477-782 77487-918
2-Chloro-2,2-difluoro-1-(2-fluoro-5-methylphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-fluoro-5-methylphenyl)ethanone

Precaución: For research use only

Formula: C₉H₆ClF₃O
MW: 222.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045708
CAS Number: 1565720-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR