2-Iodo-1-isopropoxy-4-(trifluoromethyl)benzene ≥96%

Supplier: AOBChem USA

Warning

149031-1G 149031-500MG 149031-5G 149031-25G
77465-006EA 232.78 USD
77465-006 77477-776 77487-908 77498-608
2-Iodo-1-isopropoxy-4-(trifluoromethyl)benzene ≥96%
2-Iodo-1-isopropoxy-4-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₀H₁₀F₃IO
Storage Temperature: Ambient
MDL Number: MFCD29095490
CAS Number: 1369872-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR