2-Cyano-5-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14876-1G 14876-500MG 14876-5G
77465-002EA 477.92 USD
77465-002 77477-772 77487-902
2-Cyano-5-formylphenylboronic acid ≥97%
2-Cyano-5-formylphenylboronic acid

Precaución: For research use only

Formula: C₈H₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD10697411
CAS Number: 918413-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR