3-Acetyl-5-chlorothiophene-2-boronic acid ≥97%

Supplier: AOBChem USA

Warning

14869-1G 14869-500MG 14869-5G 14869-25G
77464-998EA 106.09 USD
77464-998 77477-770 77487-898 77498-600
3-Acetyl-5-chlorothiophene-2-boronic acid ≥97%
3-Acetyl-5-chlorothiophene-2-boronic acid

Precaución: For research use only

Formula: C₆H₆BClO₃S
Storage Temperature: Ambient
MDL Number: MFCD11044926
CAS Number: 1451390-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR