2-Bromo-4-chloro-1-phenoxybenzene ≥95%

Supplier: AOBChem USA

Warning

148288-1G 148288-500MG 148288-5G 148288-25G
77464-994EA 415.09 USD
77464-994 77477-766 77487-894 77498-596
2-Bromo-4-chloro-1-phenoxybenzene ≥95%
2-Bromo-4-chloro-1-phenoxybenzene

Precaución: For research use only

Formula: C₁₂H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD34839967
CAS Number: 2378336-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR