(6-Bromo-2-fluoro-3-(methylthio)phenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148066-1G 148066-500MG 148066-5G 148066-25G
77464-990EA 415.09 USD
77464-990 77477-762 77487-890 77498-592
(6-Bromo-2-fluoro-3-(methylthio)phenyl)(furan-2-yl)methanol ≥95%
(6-Bromo-2-fluoro-3-(methylthio)phenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₀BrFO₂S
MW: 317.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839769
CAS Number: 2921849-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR