(2-Bromo-6-fluoro-4-methylphenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148051-1G 148051-500MG 148051-5G 148051-25G
77464-986EA 415.09 USD
77464-986 77477-758 77487-886 77498-588
(2-Bromo-6-fluoro-4-methylphenyl)(furan-2-yl)methanol ≥95%
(2-Bromo-6-fluoro-4-methylphenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34839754
CAS Number: 2921723-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR