(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148048-1G 148048-500MG 148048-5G 148048-25G
77464-982EA 415.09 USD
77464-982 77477-754 77487-882 77498-584
(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(furan-2-yl)methanol ≥95%
(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₈H₁₄BrFO₃
MW: 377.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839751
CAS Number: 2921692-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR