(2-Bromo-3-chloro-6-fluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148045-1G 148045-500MG 148045-5G 148045-25G
77464-978EA 415.09 USD
77464-978 77477-750 77487-878 77498-580
(2-Bromo-3-chloro-6-fluorophenyl)(furan-2-yl)methanol ≥95%
(2-Bromo-3-chloro-6-fluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₇BrClFO₂
MW: 305.527 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839748
CAS Number: 2921900-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR