(2-Chloro-6-fluoro-3-methylphenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148012-1G 148012-500MG 148012-5G 148012-25G
77464-974EA 415.09 USD
77464-974 77477-746 77487-874 77498-576
(2-Chloro-6-fluoro-3-methylphenyl)(furan-2-yl)methanol ≥95%
(2-Chloro-6-fluoro-3-methylphenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₀ClFO₂
MW: 240.658 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839716
CAS Number: 2921742-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR