(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

147992-1G 147992-500MG 147992-5G 147992-25G
77464-970EA 415.09 USD
77464-970 77477-742 77487-870 77498-572
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(furan-2-yl)methanol ≥95%
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₇BrF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34839697
CAS Number: 2921912-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR