(5-Chloro-2-fluoro-3-methylphenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

147990-1G 147990-500MG 147990-5G 147990-25G
77464-966EA 415.09 USD
77464-966 77477-738 77487-866 77498-568
(5-Chloro-2-fluoro-3-methylphenyl)(furan-2-yl)methanol ≥95%
(5-Chloro-2-fluoro-3-methylphenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34839695
CAS Number: 2921694-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR