Ethyl 2-(4-cyanophenyl)-2-hydroxyacetate ≥97%

Supplier: AOBChem USA

Warning

147883-1G 147883-500MG 147883-5G 147883-25G
77464-962EA 275.01 USD
77464-962 77477-734 77487-862 77498-564
Ethyl 2-(4-cyanophenyl)-2-hydroxyacetate ≥97%
Ethyl 2-(4-cyanophenyl)-2-hydroxyacetate

Precaución: For research use only

Formula: C₁₁H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD04038800
CAS Number: 847227-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR