5-Bromo-3,4'-bipyridine ≥95%

Supplier: AOBChem USA

Warning

147698-1G 147698-500MG 147698-5G 147698-25G
77464-958EA 168.92 USD
77464-958 77477-730 77487-858 77498-560
5-Bromo-3,4'-bipyridine ≥95%
5-Bromo-3,4'-bipyridine

Precaución: For research use only

Formula: C₁₀H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09908008
CAS Number: 106047-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR