(5-Methoxybenzo[b]thiophen-2-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

14721-1G 14721-500MG 14721-5G
77464-954EA 518.09 USD
77464-954 77477-726 77487-854
(5-Methoxybenzo[b]thiophen-2-yl)boronic acid ≥95%
(5-Methoxybenzo[b]thiophen-2-yl)boronic acid

Precaución: For research use only

Formula: C₉H₉BO₃S
MW: 208.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09952074
CAS Number: 193965-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR