1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

147060-1G 147060-500MG 147060-5G 147060-25G
77464-950EA 397.58 USD
77464-950 77477-722 77487-850 77498-554
1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)cycloheptanol ≥95%
1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)cycloheptanol

Precaución: For research use only

Formula: C₁₅H₂₀O₃
MW: 248.318 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17535667
CAS Number: 1467212-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR