3-Fluoro-2-(trimethylsilyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

146748-1G 146748-500MG 146748-5G 146748-25G
77464-946EA 415.09 USD
77464-946 77477-718 77487-846 77498-550
3-Fluoro-2-(trimethylsilyl)benzaldehyde ≥95%
3-Fluoro-2-(trimethylsilyl)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₃FOSi
Storage Temperature: Ambient
MDL Number: MFCD24682652
CAS Number: 202744-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR