2-(4-Methoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

146653-1G 146653-500MG 146653-5G
77464-942EA 1010.43 USD
77464-942 77477-716 77487-842
2-(4-Methoxyphenyl)cyclopentanol ≥95%
2-(4-Methoxyphenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₆O₂
MW: 192.2542 g/mol
Storage Temperature: Ambient
CAS Number: 933674-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR