3-Fluoro-4-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Fluoro-4-(trifluoromethoxy)phenyl)boronic acid

14550-1G 14550-5G 14550-25G
77464-934EA 64.89 USD
77464-934 77487-834 77498-538
3-Fluoro-4-(trifluoromethoxy)phenylboronic acid ≥97%
3-Fluoro-4-(trifluoromethoxy)phenylboronic acid

Precaución: For research use only

Formula: C₇H₅BF₄O₃
Storage Temperature: Ambient
MDL Number: MFCD07368680
CAS Number: 187804-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR