(E)-1-(3,5-dichlorophenyl)but-2-en-1-ol ≥97%

Supplier: AOBChem USA

Warning

145309-1G 145309-500MG 145309-5G
77464-926EA 447.02 USD
77464-926 77477-702 77487-826
(E)-1-(3,5-dichlorophenyl)but-2-en-1-ol ≥97%
(E)-1-(3,5-dichlorophenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₁₀Cl₂O
MW: 217.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30678769
CAS Number: 2635957-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR