1-(m-tolyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

145302-1G 145302-500MG 145302-5G 145302-25G
77464-922EA 415.09 USD
77464-922 77477-698 77487-822 77498-532
1-(m-tolyl)but-2-en-1-ol ≥95%
1-(m-tolyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄O
MW: 162.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30171553
CAS Number: 1195529-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR