(E)-1-(4-Bromophenyl)but-2-en-1-ol ≥97%

Supplier: AOBChem USA

Warning

145300-1G 145300-500MG 145300-5G
77464-918EA 447.02 USD
77464-918 77477-694 77487-818
(E)-1-(4-Bromophenyl)but-2-en-1-ol ≥97%
(E)-1-(4-Bromophenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₁₁BrO
MW: 227.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30678671
CAS Number: 54450-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR