(E)-1-(3-Bromophenyl)but-2-en-1-ol ≥97%

Supplier: AOBChem USA

Warning

145298-1G 145298-500MG 145298-5G
77464-914EA 447.02 USD
77464-914 77477-690 77487-814
(E)-1-(3-Bromophenyl)but-2-en-1-ol ≥97%
(E)-1-(3-Bromophenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₁₁BrO
MW: 227.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30678670
CAS Number: 54372-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR