4-Formyl-3-nitrobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

145282-1G 145282-500MG 145282-5G 145282-25G
77464-910EA 77.25 USD
77464-910 77477-686 77487-810 77498-528
4-Formyl-3-nitrobenzonitrile ≥95%
4-Formyl-3-nitrobenzonitrile

Precaución: For research use only

Formula: C₈H₄N₂O₃
MW: 176.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696869
CAS Number: 90178-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR