2-(Methoxymethoxy)benzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

14509-1G 14509-500MG 14509-5G 14509-25G
77464-906EA 138.02 USD
77464-906 77477-682 77487-806 77498-524
2-(Methoxymethoxy)benzotrifluoride ≥95%
2-(Methoxymethoxy)benzotrifluoride

Precaución: For research use only

Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09750049
CAS Number: 336628-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR