2-Bromo-6-fluorobenzene-1,4-diol ≥95%

Supplier: AOBChem USA

Warning

144588-1G 144588-500MG 144588-5G
77464-902EA 630.36 USD
77464-902 77477-678 77487-800
2-Bromo-6-fluorobenzene-1,4-diol ≥95%
2-Bromo-6-fluorobenzene-1,4-diol

Precaución: For research use only

Formula: C₆H₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28527817
CAS Number: 1785629-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR