1-Bromo-2-(2-methoxyethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

143382-1G 143382-500MG 143382-5G 143382-25G
77464-890EA 75.19 USD
77464-890 77477-666 77487-788 77498-504
1-Bromo-2-(2-methoxyethoxy)benzene ≥95%
1-Bromo-2-(2-methoxyethoxy)benzene

Precaución: For research use only

Formula: C₉H₁₁BrO₂
MW: 231.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09934824
CAS Number: 109417-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR