5-Hydroxy-2,4-dimethoxybenzaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
142694-1G
142694-500MG
142694-5G
77464-886EA
1175.23
USD
77464-886
77477-662
77487-784
5-Hydroxy-2,4-dimethoxybenzaldehyde ≥95%
5-Hydroxy-2,4-dimethoxybenzaldehyde
Precaución: For research use only
Formula:
C₉H₁₀O₄ MW: 182.173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24675799 CAS Number: 80832-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...