(3-Isopropyl-5-(methylthio)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

142298-1G 142298-500MG 142298-5G 142298-25G
77464-882EA 415.09 USD
77464-882 77477-658 77487-776 77498-492
(3-Isopropyl-5-(methylthio)phenyl)trimethylsilane ≥95%
(3-Isopropyl-5-(methylthio)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₃H₂₂SSi
Storage Temperature: Ambient
MDL Number: MFCD34835711
CAS Number: 2921842-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR