2-[2-Bromo-4-(trifluoromethyl)phenyl]-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

14202-1G 14202-500MG 14202-5G 14202-25G
77464-878EA 143.17 USD
77464-878 77477-654 77487-772 77498-488
2-[2-Bromo-4-(trifluoromethyl)phenyl]-1,3-dioxolane ≥95%
2-[2-Bromo-4-(trifluoromethyl)phenyl]-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619357
CAS Number: 1619256-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR