1-Bromo-2-((4-methoxybenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

141326-1G 141326-500MG 141326-5G
77464-874EA 457.32 USD
77464-874 77477-650 77487-768
1-Bromo-2-((4-methoxybenzyl)oxy)benzene ≥97%
1-Bromo-2-((4-methoxybenzyl)oxy)benzene

Precaución: For research use only

Formula: C₁₄H₁₃BrO₂
MW: 293.1558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11127634
CAS Number: 161886-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR