3-isopropoxy-4-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

140692-1G 140692-500MG 140692-5G
77464-870EA 836.36 USD
77464-870 77477-646 77487-764
3-isopropoxy-4-(trifluoromethyl)benzaldehyde ≥95%
3-isopropoxy-4-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₁F₃O₂
MW: 232.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29122465
CAS Number: 1289009-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR