(2-Ethoxy-4-methoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

140368-1G 140368-500MG 140368-5G
77464-866EA 477.92 USD
77464-866 77477-642 77487-760
(2-Ethoxy-4-methoxyphenyl)boronic acid ≥95%
(2-Ethoxy-4-methoxyphenyl)boronic acid

Precaución: For research use only

Formula: C₉H₁₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD22414601
CAS Number: 2898529-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR