1-(2,5-dimethylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140311-1G 140311-500MG 140311-5G 140311-25G
77464-862EA 260.59 USD
77464-862 77477-640 77487-756 77498-482
1-(2,5-dimethylphenyl)-2-methylpropan-1-ol ≥95%
1-(2,5-dimethylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O
MW: 178.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11934822
CAS Number: 139448-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR