1-(4-Methoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140282-1G 140282-500MG 140282-5G
77464-858EA 546.93 USD
77464-858 77477-636 77487-752
1-(4-Methoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Methoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24363301
CAS Number: 1556459-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR