1-(2,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140215-1G 140215-500MG 140215-5G 140215-25G
77464-854EA 415.09 USD
77464-854 77477-632 77487-748 77498-476
1-(2,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2,5-dimethylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD12787272
CAS Number: 66390-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR