1-(4-butylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140202-1G 140202-500MG 140202-5G 140202-25G
77464-850EA 397.58 USD
77464-850 77477-628 77487-744 77498-472
1-(4-butylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-butylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787353
CAS Number: 1284094-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR